Learn More
Kimberly-Clark Professional™ KleenGuard™ A20 Breathable Particle Protection Coveralls: Open Wrists and Ankles
GREENER_CHOICE
Patented Reflex™ design is 7.5 times less likely to rip out than ANSI™ minimums — provides 12% more chest room and 6% longer body length
Manufacturer:  Kimberly-Clark Professional™ 49004
 View more versions of this product
Catalog No. 17-981-44B
  • $36.88 / Case of 24
    Save 84%
    Reg : $226.00
    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
    Learn More
Add to cart
Alert:
Not for use near heat, flames or flammable environments
This item is not returnable. View return policy
Description
Description
For use in laboratories, remediation, decontamination modesty garment, pesticides, and heavy dust environments. With open wrists and ankles.
  • Provide general protection against many particles
  • Breathable patented Microforce™ barrier SMS fabric passes NFPA 99 for antistatic materials
  • Offers high abrasion resistance and is low linting
  • Withstand multiple wearings
  • Air and moisture vapor can pass through to aid wearer comfort
  • Triple-stitched serged seams
  • White
Specifications
Specifications
X-Large
White
Open
NFPA 99
Laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments
SMS
Open
Front zipper
Microforce™ Barrier SMS Fabric