Learn More
Kimberly-Clark Professional™ KleenGuard™ A20 Breathable Particle Protection Denim Blue Coveralls Available on GSA/VA Contract for Federal Government customers only.
Strong, breathable, abrasion-resistant fabric keep workers cool and comfortable while protecting against fibers, fine dusts, and low surface tension liquid splashes
Manufacturer:  Kimberly-Clark Professional™ 58524
 View more versions of this product
Catalog No. 19-057-401C
Add to cart
Description
Description
Patented Reflex™ design is 7.5 times less likely to rip out than ANSI™ minimums — provides 12% more chest room and 6% longer body length.
  • Ideal for use in dusty environments, as well as for painting, manufacture spraying, and agricultural spraying
  • Stylish denim blue color masks dirt and grease
  • Air and moisture vapor can pass through to aid wearer comfort
  • Triple-stitched serged seams
  • Can withstand multiple wearing
  • Zippered front
  • Attached hood and boots
  • Elastic back, wrists and ankles
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Attached boots, Elastic
Elastic
Zipper front
Elastic
SMS Fabric
Serged
Safety
ANSI, NFPA 99
Denim Blue
Laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments
Hooded
X-Large