Learn More
Kimberly-Clark™ Professional KleenGuard™ A20 Breathable Particle Protection Denim Blue Coveralls
Strong, breathable, abrasion-resistant fabric keep workers cool and comfortable while protecting against fibers, fine dusts, and low surface tension liquid splashes
Manufacturer:  Kimberly-Clark™ Professional 58532
 View more versions of this product
Catalog No. 19-057-400A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ANSI, NFPA 99
Microforce™ Barrier SMS Fabric
Open
Serged
Medium
Zipper Front
Open
SMS Fabric
Denim Blue
Laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments