Learn More
Kimberly-Clark™ Professional KleenGuard™ A20 Breathable Particle Protection Denim Blue Coveralls Available on GSA/VA Contract for Federal Government customers only.
Strong, breathable, abrasion-resistant fabric keep workers cool and comfortable while protecting against fibers, fine dusts and low surface tension liquid splashes
Manufacturer:  Kimberly-Clark™ Professional 58502
 View more versions of this product
Catalog No. 19-088-1929A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ANSI, NFPA 99
Elastic
Elastic
SMS Fabric
Denim Blue
Laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments
Zipper Front
Microforce™ Barrier SMS Fabric
Elastic
Serged
Medium