Kimberly-Clark™ Professional KleenGuard™ A20 Breathable Particle Protection Denim Blue Coveralls Available on GSA/VA Contract for Federal Government customers only.

Strong, breathable, abrasion-resistant fabric keep workers cool and comfortable while protecting against fibers, fine dusts and low surface tension liquid splashes

Manufacturer: Kimberly-Clark™ Professional 58503

 View more versions of this product

Catalog No. 19-088-1929B

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ANSI, NFPA 99
    Material SMS Fabric
    Seam Type Serged
    Color Denim Blue
    Size Large
    For Use With (Application) Laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments
    Closure Type Zipper Front
    Back Style Elastic
    Construction Microforce™ Barrier SMS Fabric
    Ankle Style Elastic
    Cuff Style Elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit