Kimberly-Clark™ Professional KleenGuard™ A20 Lab Coat with Knit Cuffs Available on GSA/VA Contract for Federal Government customers only.

Breathable particle protection with comfortable snug fit

Manufacturer: Kimberly-Clark™ Professional 23875

 View more versions of this product

Catalog No. 19-020-2500

  Check Availability
Add to Cart

    Specifications

    Material Microforce™ barrier SMS Fabric
    No. of Pockets 2.0
    Size 2X-Large
    For Use With (Application) Suited for laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments
    Collar Type High mandarin
    Certifications/Compliance NFPA 99
    Closure Type Plastic Snap Front
    Color Denim Blue
    Length (English) 46 in.
    Cuff Style Knit
    Length (Metric) 117cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit