Learn More
Kimberly-Clark Professional™ KleenGuard™ A30 Breathable Splash Particle Protection Coveralls with Elastic Back Get IT Item
Combine durability and breathability to provide comfortable splash and particle protection
Manufacturer:  Kimberly-Clark Professional™ 46005
 View more versions of this product
Catalog No. 19-170-386D
Add to cart
Description
Description
MICROFORCE™ Barrier SMS Fabric incorporates a web of microfibers that filter particulates and water-based liquids. Suitable for many applications including pharmaceutical research, food processing and more.
  • Excellent protection from nonhazardous particles and liquids
  • Strong and abrasion resistant; suitable for multiple wearings
  • Seamless front to provide more protection in the primary exposure areas
  • Zipper front with 1 in. (2.5cm) flap and elastic back
  • Color: White
  • With elastic back
Compliance
Passes NFPA 99 for antistatic materials.
Specifications
Specifications
Elastic
White
Open
Open
2X-Large
Polypropylene
Documents
Accessories