Learn More
Kimberly-Clark Professional™ KleenGuard™ A30 Breathable Splash Particle Protection Coveralls with Elastic Back
GREENER_CHOICE
Combine durability and breathability to provide comfortable splash and particle protection
$363.00 - $381.00
Specifications
Material Polypropylene
Color White
Ankle Style Open
Cuff Style Open
Back Style Elastic
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-170-386E
GREENER_CHOICE
Accessories View Documents
Kimberly-Clark Professional™
46006
3X-Large
Case of 21 for $363.00
Add to cart
19-170-386B
GREENER_CHOICE
Accessories View Documents
Kimberly-Clark Professional™
46003
Large
Case of 25 for $381.00
Add to cart
19-170-386D
GREENER_CHOICE
Accessories View Documents
Kimberly-Clark Professional™
46005
2X-Large
Case of 25 for $381.00
Add to cart
19-170-386A
GREENER_CHOICE
Accessories View Documents
Kimberly-Clark Professional™
46002
Medium
Case of 25 for $381.00
Add to cart
19-170-386C
GREENER_CHOICE
Accessories View Documents
Kimberly-Clark Professional™
46004
X-Large
Case of 25 for $381.00
Add to cart
19-170-386F
GREENER_CHOICE
Accessories View Documents
Kimberly-Clark Professional™
46007
4X-Large
Case of 21 for $363.00
Add to cart
Description
Description
MICROFORCE™ Barrier SMS Fabric incorporates a web of microfibers that filter particulates and water-based liquids. Suitable for many applications including pharmaceutical research, food processing and more.
  • Excellent protection from nonhazardous particles and liquids
  • Strong and abrasion resistant; suitable for multiple wearings
  • Seamless front to provide more protection in the primary exposure areas
  • Zipper front with 1 in. (2.5cm) flap and elastic back
  • Color: White
  • With elastic back
Compliance
Passes NFPA 99 for antistatic materials.
Specifications
Specifications
Polypropylene
Open
Elastic
White
Open