Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ A35 Liquid and Particle Protection Coveralls
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description:
Elastic wrists and ankles
Elastic wrists, ankles and hood
Elastic wrists, ankles, hood and boots
Non-Elastic; Zipper Front
Zipper Front, Elastic Wrists and Ankles and Hood
Description
- Breathable
- Microporous film laminate
- Lightweight outer fabric that protects the microporous film laminate
- Added durability and comfort liquid barrier
- Particulate barrier
- Available with elastic wrists and ankles
- With or without hood
- Zipper front
Specifications
Size | Large |
Material | Microporous Film Laminate |
Color | White |
Disposable | Single-use |
Waist Style | Non-elastic |
Closure Type | Front Zipper |
Seam Type | Serged |
Certifications/Compliance | ANSI/ISEA 101-1996 |
Description | Non-Elastic; Zipper Front |
For Use With (Application) | Liquid handling, pressure washing, maintenance and general purpose |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction