Kimberly-Clark™ Professional KleenGuard™ A35 Liquid and Particle Protection Coveralls

Provide a comfortable fit while providing liquid and particulate protection

Manufacturer: Kimberly-Clark™ Professional 38935

 View more versions of this product

Catalog No. 19-006-218

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ANSI/ISEA 101-1996
    Material Microporous Film Laminate
    Includes Elastic wrists and ankles
    Seam Type Serged
    Color White
    Size 5XL
    Disposable Yes
    For Use With (Application) Liquid handling, pressure washing, maintenance and general purpose
    Closure Type Zipper front
    Waist Style Elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit