Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional KleenGuard A40 Liquid and Particle Protection Sleeves
Breathable, microporous laminate protects against nonhazardous liquids
$139.00
Specifications
Certifications/Compliance | NFPA 99, ASTM F1670 |
---|---|
Color | White |
Material | Tyvek™ |
Size | Universal |
For Use With (Application) | Fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance, general manufacturing |
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Price | Quantity & Availability | ||||
19-120-2406
|
Kimberly-Clark Professional
44480 |
18 in. | 45.7 cm |
Case of 200 for $139.00
|
|
Description
White, disposable sleeve protectors.
- 18 in. long (45.7cm) sleeve protectors feature elastic ends for a secure fit
- Low lint
Compliance
Meet NFPA 99 for Antistatic Materials and ASTM F1670 testing for penetration of blood and bodily fluids.
Specifications
NFPA 99, ASTM F1670 | |
Tyvek™ | |
Fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance, general manufacturing |
White | |
Universal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title