Kimberly-Clark™ Professional KleenGuard™ A40 Liquid and Particle Protection Coveralls Available on GSA/VA Contract for Federal Government customers only.
Excellent protection from nonhazardous liquids
Manufacturer: Kimberly-Clark™ Professional 44344
 View more versions of this product
Catalog No. 19-130-3477D
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications/Compliance ASTM F1670, NFPA 99, ANSI ISEA 101-1996
    Material SMS Fabric
    Includes Zipper front; Individually wrapped To-Go
    Color White
    Size X-Large
    Disposable Yes
    For Use With (Application) Fiberglass manufacturing, liquid handling, utilities, pressure washing, tank cleaning, maintenance and general manufacturing
    Closure Type Zipper Front
    Construction Non-woven Microporous film laminate barrier over SMS
  • Description & Specifications
Related Products
Product Content Feedback