Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ A40 Liquid & Particle Protection Coveralls
Click to view available options
With hood; Zipper front; Elastic wrists and ankles; XX-Large
Description
Excellent protection from nonhazardous liquids
Microporous, nonwoven film laminate offers an improved liquid barrier over SMS apparel.
- Low linting
- White
- Zipper front
Compliance
Meets ANSI™/ISEA™ 101-1996; ASTM™ F1670 testing for penetration of blood and bodily fluids; NFPA 99 Health Care Facilities criteria for antistatic materials.
Specifications
Size | 2X-Large |
Material | SMS |
Color | White |
Disposable | Single-use |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | Elastic |
Closure Type | Front Zipper |
Back Style | Breathable |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction