Learn More
Kimberly-Clark™ Professional KleenGuard™ A40 Liquid and Particle Protection Coveralls
Excellent protection from nonhazardous liquids
Manufacturer:  Kimberly-Clark™ Professional 44322
 View more versions of this product
Catalog No. 19-120-2400B
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ASTM F1670, NFPA 99, ANSI ISEA 101-1996
Non-woven Microporous film laminate barrier over SMS
Elastic
Zipper Front, Elastic Wrist, Ankles and Hood
Medium
Fiberglass manufacturing, liquid handling, utilities, pressure washing, tank cleaning, maintenance and general manufacturing
Zipper Front
Elastic
Polypropylene
White
Yes
Accessories