Learn More
Kimberly-Clark™ Professional KleenGuard™ A40 Liquid and Particle Protection Lab Coats Get IT Item Available on GSA/VA Contract for Federal Government customers only.
Provides better liquid and particulate barrier
Manufacturer:  Kimberly-Clark™ Professional 44446
 View more versions of this product
Catalog No. 19-120-2403F
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Traditional
Snap
Elastic
3X-Large
No pockets and elastic wrists
NFPA 99, ASTM F1670, ANSI ISEA 101-1996
White
Polypropylene
Suited for Fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance and general manufacturing
Microporous Film Laminate