Kimberly-Clark™ Professional KleenGuard™ A40 Liquid and Particle Protection Lab Coats Available on GSA/VA Contract for Federal Government customers only.
Provides better liquid and particulate barrier
Manufacturer: Kimberly-Clark™ Professional 44454
 View more versions of this product
Catalog No. 19-120-2404D
  Check Availability
Add to Cart
    Specifications
    Product Type Chest and hip pockets
    No. of Pockets 2.0
    Construction Microporous Film Laminate
    Cuff Style Elastic
    Material Polypropylene
    Size X-Large
    For Use With (Application) Suited for Fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance and general manufacturing
    Collar Type Traditional
    Certifications/Compliance NFPA 99, ASTM F1670, ANSI ISEA 101-1996
    Closure Type Snap Front
    Color White
  • Description & Specifications
Related Products
Product Content Feedback