Learn More
Kimberly-Clark™ Professional KleenGuard™ A40 Liquid and Particle Protection Lab Coats Available on GSA/VA Contract for Federal Government customers only.
Provides better liquid and particulate barrier
Manufacturer:  Kimberly-Clark™ Professional 44455
 View more versions of this product
Catalog No. 19-120-2404E
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Traditional
Snap
Elastic
2
Suited for Fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance and general manufacturing
Microporous Film Laminate
NFPA 99, ASTM F1670, ANSI ISEA 101-1996
White
Polypropylene
2X-Large
Chest and hip pockets