Kimberly-Clark™ Professional KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Coveralls Available on GSA/VA Contract for Federal Government customers only.
Provide exceptional protection against nonhazardous liquids, dry particulates, and bloodborne pathogens, even under pressure
Manufacturer: Kimberly-Clark™ Professional 45027
 View more versions of this product
Catalog No. 19-057-404F
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance ASTM F1670, 1671
    Closure Type Zipper Front
    Construction 3-ply Fabric construction middle layer of Microporous film
    Material Fabric
    Includes Hooded, seamless zipper front with storm flap, elastic wrist and ankle, elastic across the back
    Color Denim Blue
    Size 4X-Large
    For Use With (Application) Animal research, emergency medical response, crime scene cleanup, embalming/forensics, tank cleaning, Fiberglass manufacturing, liquid handling, food processing
    Resistance Bloodborne pathogen and chemical
    Breathable fabric is strong, comfortable, and abrasion-resistant, with a three-layer construction designed to allow heat and sweat vapor to escape.
    • Patented Reflex™ Design is 7.5 times less likely to rip out than ANSI™ minimum standards
    • Feature 12% more chest room and 6% longer body length
    • Provide excellent front and strikethrough protection with a seamless front and concealed 28 in. (71cm) zipper
    • Used in many industries and applications, including emergency medical, liquid handling, forensics, nuclear facilities and more
    • Denim blue
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback