Kimberly-Clark™ Professional KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Coveralls Available on GSA/VA Contract for Federal Government customers only.
Provide exceptional protection against nonhazardous liquids, dry particulates, and bloodborne pathogens, even under pressure
Manufacturer: Kimberly-Clark™ Professional 45232
 View more versions of this product
Catalog No. 19-057-402A
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance ASTM F1670, 1671
    Material Fabric
    Includes Seamless zipper front, open wrist and ankle, Reflex™ design
    Color Denim Blue
    Size Medium
    For Use With (Application) Animal research, emergency medical response, crime scene cleanup, embalming/forensics, tank cleaning, Fiberglass manufacturing, liquid handling, food processing
    Resistance Bloodborne pathogen and chemical
    Closure Type Zipper Front
    Construction 3-ply Fabric construction middle layer of Microporous film
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback