Learn More
Kimberly-Clark Professional™ KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Coveralls Available on GSA/VA Contract for Federal Government customers only.
Provide exceptional protection against nonhazardous liquids, dry particulates, and bloodborne pathogens, even under pressure
Manufacturer:  Kimberly-Clark Professional™ 45022
 View more versions of this product
Catalog No. 19-057-404A
Add to cart
Description
Description
Breathable fabric is strong, comfortable, and abrasion-resistant, with a three-layer construction designed to allow heat and sweat vapor to escape.
  • Patented Reflex™ Design is 7.5 times less likely to rip out than ANSI™ minimum standards
  • Feature 12% more chest room and 6% longer body length
  • Provide excellent front and strikethrough protection with a seamless front and concealed 28 in. (71cm) zipper
  • Used in many industries and applications, including emergency medical, liquid handling, forensics, nuclear facilities and more
  • Denim blue
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Compliance
Meet ASTM F1670/1671 specifications for blood, bodily fluids and bloodborne pathogens.
Specifications
Specifications
Elastic
Elastic
Zipper front
Elastic
Animal research, emergency medical response, crime scene cleanup, embalming/forensics, tank cleaning, Fiberglass manufacturing, liquid handling, food processing
Hooded
Safety
ASTM F1670, 1671
Denim Blue
Hooded, seamless zipper front with storm flap, elastic wrist and ankle, elastic across the back; 3-ply Fabric construction middle layer of Microporous film
Fabric
Medium
Documents
Accessories