Learn More
Kimberly-Clark Professional™ KleenGuard™ A65 Flame-Resistant Coveralls
Soft, lightweight and breathable
Manufacturer:  Kimberly-Clark Professional™ 45316
 View more versions of this product
Catalog No. 19-057-457E
Add to cart
Description
Description
Designed to be worn in areas with potential exposure to sparks, these coveralls are made of a comfortable, breathable and durable fabric treated with a flame-resistant finish.
  • Design meets ANSI™/ISEA™ 101-1996 sizing standard
  • Ideal for welding, oil refining, utilities, grinding and general maintenance applications
  • Zipper front
Compliance
While not intended as primary fire protection apparel, the fabric is flame-resistant and compliant with NFPA™ 701-1989 Flame Propagation test, meaning it will not melt or propagate a flame.
Specifications
Specifications
Open
ANSI ISEA 101-1996, CAN CGSB-155.20-2000, NFPA 701, NFPA 99
Blue
Durable, spunlace, non-woven Fabric treated with flame-resistant finish
37.75 in.
Fabric
54 to 56 in.
56 in.
81.28cm
Safety
Zipper front
Open
Welding, Oil refining, utilities, grinding
14.86cm
3X-Large
32 in.
137.16 to 142.24cm
142.24cm