Learn More
Kimberly-Clark™ Professional KleenGuard™ A65 Flame-Resistant Coveralls Available on GSA/VA Contract for Federal Government customers only.
Soft, lightweight and breathable
Manufacturer:  Kimberly-Clark™ Professional 45354
 View more versions of this product
Catalog No. 19-165-282D
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ANSI ISEA 101-1996, CAN CGSB-155.20-2000, NFPA 701, NFPA 99
Durable, spunlace, non-woven Fabric treated with flame-resistant finish
Elastic
Blue
46 to 48 in.
36.25 in.
32 in.
81.28cm
Zipper Front
Elastic
Fabric
X-Large
Welding, Oil refining, utilities, grinding
14.27cm
42 in.
106.68cm