Kimberly-Clark™ Professional KleenGuard™ A65 Flame-Resistant Lab Coats Available on GSA/VA Contract for Federal Government customers only.

Designed for use in areas where exposure to sparks or flame is possible

Manufacturer: Kimberly-Clark™ Professional 12887

 View all options for this product

Catalog No. 19-047-728


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Certifications/Compliance NFPA 701, CAN CGSB-155.20-2000, NFPA 99
    Closure Type Hook and loop
    Color Blue
    Material Fabric
    Size 6X-Large
    For Use With (Application) Ideal for use in laboratories, use in areas where exposure to sparks or flame is possible
    Resistance Flame, tear

    Ideally suited for use in laboratories.

    • Comfortable, breathable and durable fabric resists tearing
    • Hook and loop closures
    • No pocket
    • May also be used to protect primary fire protection clothing

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit