Learn More
Kimberly-Clark Professional™ KleenGuard™ A65 Flame-Resistant Lab Coats
Designed for use in areas where exposure to sparks or flame is possible
Manufacturer:  Kimberly-Clark Professional™ 12815
 View more versions of this product
Catalog No. 19-047-725
Add to cart
Description
Description
Ideally suited for use in laboratories.
  • Comfortable, breathable and durable fabric resists tearing
  • Hook and loop closures
  • No pocket
  • May also be used to protect primary fire protection clothing
Compliance
Meets NFPA 701 FLAME PROPAGATION TEST, 1989 Edition. Complies with the flame-resistance requirements cited by paragraph 6.1 of CAN/CGSB-155.20-2000. NFPA 99 compliant antistatic material.
Specifications
Specifications
Safety
Hook and loop
Ideal for use in laboratories, use in areas where exposure to sparks or flame is possible
4X-Large
58 to 60 in.
7 oz.
76.2cm
NFPA 701, CAN CGSB-155.20-2000, NFPA 99
Blue
Polyester
Flame, tear
147.32 to 152.4 cm
0.19 kg
30 in.