Learn More
Kimberly-Clark Professional™ KleenGuard™ A70 Chemical Spray Coveralls Available on GSA/VA Contract for Federal Government customers only.
Reflex™ design offers 12% more chest room and 6% longer body length
Manufacturer:  Kimberly-Clark Professional™ 00687
 View more versions of this product
Catalog No. 19-166-490
Add to cart
Description
Description
Coveralls provide protection for hazardous waste remediation, environmental cleanup, petrochemical and oil refining, acid and caustic handling, biohazard cleanup, tank cleaning and pharmaceutical manufacturing.
  • Feature bound seams
  • Storm flap provides a seamless chest area
  • Water-resistant zipper and wick-away lining for enhanced comfort
  • Yellow
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Compliance
Meet ASTM™ F 903 penetration test standards.
Specifications
Specifications
ASTM F 903, NFPA 99
Bound
4X-Large
1.5mil
Elastic
Hooded, zipper front, storm flap, elastic wrist and ankle, Reflex™ design, wick-away lininig; 1.5 mil Polyethylene film Coated Spunbond Polyproylene Fabric
Elastic
Yellow
Hazardous waste remediation, environmental cleanup, petrochemical refining, Acid/Caustic handling, biohazard cleanup, tank cleaning, pharmaceutical manufacturing
Zipper Front and Storm Flap
Hooded
Polyethylene, Spunbond Polyproylene Fabric