Learn More
Kimberly-Clark Professional™ KleenGuard™ A70 Chemical Spray Coveralls
Reflex™ design offers 12% more chest room and 6% longer body length
Manufacturer:  Kimberly-Clark Professional™ 09813
 View more versions of this product
Catalog No. 19-166-492
Add to cart
Description
Description
Coveralls provide protection for hazardous waste remediation, environmental cleanup, petrochemical and oil refining, acid and caustic handling, biohazard cleanup, tank cleaning and pharmaceutical manufacturing.
  • Feature bound seams
  • Storm flap provides a seamless chest area
  • Water-resistant zipper and wick-away lining for enhanced comfort
  • Yellow
Compliance
Meet ASTM™ F 903 penetration test standards.
Specifications
Specifications
ASTM F 903, NFPA 99
Bound
Large
1.5mil
Elastic
Hooded, zipper front, storm flap, elastic wrist and ankle, Reflex™ design, wick-away lininig; 1.5 mil Polyethylene film Coated Spunbond Polyproylene Fabric
Elastic
Yellow
Hazardous waste remediation, environmental cleanup, petrochemical refining, Acid/Caustic handling, biohazard cleanup, tank cleaning, pharmaceutical manufacturing
Zipper Front and Storm Flap
Hooded
Polyethylene, Spunbond Polyproylene Fabric