Kimberly-Clark™ Professional KleenGuard™ A70 Chemical Spray Coveralls Available on GSA/VA Contract for Federal Government customers only.
Reflex™ design offers 12% more chest room and 6% longer body length
Manufacturer: Kimberly-Clark™ Professional 09814
 View more versions of this product
Catalog No. 19-166-493
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance ASTM F 903, NFPA 99
    Material Polyethylene, Spunbond Polyproylene Fabric
    Includes Hooded and booted, zipper front, storm flap, elastic wrist, Reflex™ design, wick-away lininig
    Seam Type Bound
    Color Yellow
    Size X-Large
    For Use With (Application) Hazardous waste remediation, environmental cleanup, petrochemical refining, Acid/Caustic handling, biohazard cleanup, tank cleaning, pharmaceutical manufacturing
    Resistance Chemical
    Closure Type Zipper Front and Storm Flap
    Thickness 1.5mil
    Construction 1.5 mil Polyethylene film Coated Spunbond Polyproylene Fabric
  • Description & Specifications
Related Products
Product Content Feedback