Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

Kimberly-Clark™ Professional KleenGuard™ G10 Arctic Blue Nitrile Gloves

Provide the protection, performance and comfort of nitrile at an affordable everyday latex price

Manufacturer:  Kimberly-Clark™ Professional 90099

 View more versions of this product

Catalog No. 19-021-004



 
Add to cart

Specifications Description & Specifications

Specifications

Textured fingertips
Yes
Ambidextrous
Yes
X-Large
12cm
21 CFR 170-199
Arctic Blue
9.5 in.
9.5 x 4.72 in. (24 x 12cm)
Beaded/Rolled
Industrial, Food
Nitrile
4.72 in.
2mil
Powder-free
Non-sterile
24cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit