Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark™ Professional KleenGuard™ G10 Gray Nitrile Gloves

Designed for tasks requiring maximum dexterity

Manufacturer:  Kimberly-Clark™ Professional 97822

 View more versions of this product

Catalog No. 19-169-970C



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Textured fingertips
Beaded/Rolled
Industrial
Nitrile
3.74 in.
3.5mil
Gray
9.5 in.
9.5 x 3.74 in. (24 x 9.5cm)
Ambidextrous
Yes
Medium
9.5cm
Powder-free
Non-sterile
24cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit