Learn More
Kimberly-Clark™ Professional Scott™ Clear Antibacterial Skin Cleanser Available on GSA/VA Contract for Federal Government customers only.
Mild, general-purpose antibacterial cleanser designed for everyday use
Manufacturer:  Kimberly-Clark™ Professional 91440
 View more versions of this product
Catalog No. 19-412-302
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Clear
For everyday use
Antibacterial Skin Cleanser
1000mL
Triclosan 0.25%, Aloe Vera, emollients
Light floral
34 oz.
3 Case