Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark™ Professional Scott™ Clear Antibacterial Skin Cleanser

Mild, general-purpose antibacterial cleanser designed for everyday use

Specifications

Color Clear
Product Type Antibacterial Skin Cleanser
For Use With (Application) For everyday use
View More Specs

Products 1
Catalog Number Mfr. No. Size (English) Quantity Size (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Quantity Size (Metric) Price Quantity & Availability  
19-412-302
View Documents
Kimberly-Clark™ Professional
91440
34 oz. 3 Case 1000mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Active ingredient, 0.25% Triclosan, is effective against a wide range of bacteria
  • Helps protect against the spread of potentially harmful bacteria
  • Contains aloe vera and emollients that moisturize the skin
  • pH balanced crystal-clear formula
  • Light floral scent

Order Info

Packaged in 27 oz. (800mL) and 34 oz. (1000mL) refill. Dispenser sold separately; contact your Fisher Safety Customer Safety Representative for ordering information.

Specifications

Specifications

Clear
Antibacterial Skin Cleanser
For everyday use
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit