Learn More
Kimberly-Clark Professional™ Smith & Wesson™ Caliber™ Safety Eyewear
Precision, cut-to-shape spherical lens provides increased peripheral and distortion-free protection
Manufacturer:  Kimberly-Clark Professional™ 23007
 View more versions of this product
Catalog No. 19-706-018
Add to cart
Description
Description
  • Ergonomic, rubber nose pads with built-in airflow holes adjust to fit any nose shape
  • Scratch-resistant polycarbonate lens with antifog coating
  • Hinge stops prevent lens damage
  • Contoured, soft-touch rubber temples
  • Seven-base curve spherical lens
Compliance
Meets ANSI™ Z87.1-2010 and CSA Z94.3 standards.
Specifications
Specifications
Smoke
Safety
Black
Scratch-resistant
Meets ANSI Z87.1-2010 and CSA Z94.3 standards.
Polycarbonate