Learn More
Kimberly-Clark Professional™ Spring Green Nitrile Powder-Free Exam Gloves Get IT Item
Recyclable through RightCycle™ program
Manufacturer:  Kimberly-Clark Professional™ 43440
 View more versions of this product
Catalog No. 19-169-070
Add to cart
Description
Description
  • Material: Nitrile
  • Thickness: 4.7mil
  • Ambidextrous
  • Textured fingertips
  • Beaded cuff, wet and dry grip
  • Free of natural rubber and latex
Certifications
0.65 AQL exceeds ASTM 2.5 standard for critical defects (pinholes); 510K approved for medical use
Compliance
Meets or exceeds ASTM™ 2.5 standards; FDA 510(k) approved for medical use.
Specifications
Specifications
Beaded
Large
ASTM 2.5, FDA 510(k)
Ambidextrous
4.7 mil