Kimberly-Clark™ Professional Synthetic Plus Vinyl Exam Gloves
Economical choice for users with sensitive skin
Manufacturer: Kimberly-Clark™ Professional 50031
 View more versions of this product
Catalog No. 19-130-4852A
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Exterior Finish Smooth
    Glove Cuff Style Beaded
    Hand Ambidextrous
    Length (English) 9.5 in.
    Length (Metric) 24.1cm
    Powdered No
    Size Small
    Sterile No
    Color White
    Latex Free Yes
    Material Vinyl
    Thickness 5.1mil
  • Description & Specifications
Related Products
Product Content Feedback