Kimble™ Adapters for Round-Bottom Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Required when centrifuging select Kimble high-strength round-bottom centrifuge tubes to 13,000xG in 50mL rotor
Manufacturer: Kimble™ 4555030
 View more versions of this product
Catalog No. 05-569-11
  Check Availability
Add to Cart
    Specifications
    Material Rubber
    For Use With (Equipment) 30mL Round bottom centrifuge tube
  • Description & Specifications
Related Products
Product Content Feedback