Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
DWK Life Sciences Kimble™ Aluminum Seals for Headspace Vials
Allow for easy access to vial contents
Manufacturer:  DWK Life Sciences 7382020
 View more versions of this product
Catalog No. 03-340-27
Description
Description
  • One piece
  • Fits GPI Aluminum Seal Finish 20/20A
  • Unlined
  • Aluminum
  • 20mm (0.787 in.) size
Model 73820-20
  • Tear-out style
  • Model 73821-20
    • Tear-off style
    Specifications
    Specifications
    Aluminum
    1000 Case
    Tear-out One Piece Seal
    Tear-out
    20mm
    Natural