Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ Amber Boston Rounds with PTFE-Faced LDPE Foam-lined Caps

Narrow-mouth design provides more controlled pouring than wide-mouth.

Manufacturer:  Kimble™ 5121628V26

 View more versions of this product

Catalog No. 13-756-586


  Check Availability
Add to cart

Specifications

Description

  • Amber colored glass protects contents from UV rays and is ideal for light sensitive products.

Description & Specifications

Specifications

16 oz., 500mL
16 oz.
PTFE/LDPE
2.95 in.
Glass
Boston Round Bottles
28-400
168mm
500mL
Phenolic
Amber
6.6 in.
Convenience Pack (cap attached)
Black
75mm
Screw
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit