Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble Black with Cone-Shaped Liner Phenolic Caps/Closures
Black caps and closures offer excellent sample storage and re-sealing properties
$108.00 - $117.00
Specifications
Color | Black |
---|---|
For Use With (Application) | Excellent sample storage and re-sealing properties |
Closure Material | Phenolic |
Liner Material | Polyethylene |
Catalog Number | Mfr. No. | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Thread Size | Price | Quantity & Availability | |||||
03-340-207
|
DWK Life Sciences
7380915425 |
15-425 |
Case of 144 for $111.00
|
|
03-340-206
|
DWK Life Sciences
7380913425 |
13-425 |
Case of 144 for $108.00
|
|
03-340-208
|
DWK Life Sciences
7380918400 |
18-400 |
Case of 144 for $114.00
|
|
03-340-210
|
DWK Life Sciences
7380922400 |
22-400 |
Case of 144 for $117.00
|
|
Description
- Phenolic
- Cone-shaped liner for two-part seal
- Superior torque retention
Specifications
Black | |
Excellent sample storage and re-sealing properties | |
Phenolic | |
Polyethylene |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title