Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ 2mL and 4mL Autosampler Vials without Closures:Screw Threads

Constructed of borosilicate glass

Manufacturer:  DWK Life Sciences 331545SW

 View more versions of this product

Catalog No. 03-341-629


Add to cart

Description

Description

  • Finish: Screw thread

Specifications

Specifications

13-425
Clear
Borosilicate Glass
Yes
Screw cap
1.77 in.
4mL Autosampler Vial with marking spot
15mm
4.0mL
100
Chromatography applications
45mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit