Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ 51 Expansion Borosilicate Glass Melting Point Capillaries with Smaller I.D. Available on GSA/VA Contract for Federal Government customers only.

Manufacturer:  Kimble™ 3450799

 View more versions of this product

Catalog No. 12-141-10


  Check Availability
Add to cart

Specifications

Description

  • I.D.: 0.8 to 1.1mm
  • 100 capillaries per vial

Description & Specifications

Specifications

USP type I, ASTM E438
0.8 to 1.1mm
3.54 in.
0.25mm
Melting Point Capillary
0.03149 to 0.0433 in.
One end sealed
90mm
Borosilicate Glass
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit