Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials without Closures
Manufactured from amber 33 expansion borosilicate glass
$165.90 - $352.00
Specifications
Certifications/Compliance USP Type I
Closure Type Screw Cap
Color Amber
Material Borosilicate Glass
Thread Style Screw
View More Specs
Products 8
Catalog Number Mfr. No. Volume (English) Thread Size Price Quantity & Availability  
Catalog Number Mfr. No. Volume (English) Thread Size Price Quantity & Availability  
14-957-262
View Documents
DWK Life Sciences
609121545
1 dr. 13-425
Case of 200 for $171.20
Add to cart
14-957-265
View Documents
DWK Life Sciences
609121528
0.5 dr. 13-425
Case of 200 for $165.90
Add to cart
14-957-264
View Documents
DWK Life Sciences
609121760
2 dr. 15-425
Case of 200 for $186.90
Add to cart
14-957-266
View Documents
DWK Life Sciences
609121738
1 dr. 15-425
Case of 200 for $165.90
Add to cart
14-957-268
View Documents
DWK Life Sciences
609122857
5 dr. 24-400
Case of 200 for $291.00
Add to cart
14-957-267
View Documents
DWK Life Sciences
609121940
1.5 dr. 15-425
Case of 200 for $175.40
Add to cart
14-957-269
View Documents
DWK Life Sciences
609122895
10 dr. 24-400
Case of 200 for $352.00
Add to cart
14-957-263
View Documents
DWK Life Sciences
609121235
0.5 dr. 8-425 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Amber vials offer protection against harmful light and UV rays for light sensitive products.
Specifications
Specifications
USP Type I
Amber
Screw
Screw Cap
Borosilicate Glass