Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached Closures
SDP
Attached open top white polypropylene closures
Supplier:  DWK Life Sciences 60811S10
 View more versions of this product
Catalog No. 14-957-225
Add to Cart
Description
Description
  • Manufactured from clear 33 Expansion borosilicate glass
  • Closures supplied with PTFE-faced silicone liners
  • Vials are packaged in corrugated trays with partitions
Specifications
Specifications
A
Clear
Borosilicate Glass
Screw
Polypropylene
95 mm
PTFE/Silicone
28 mm
40 mL
10.82 dr.
Screw Cap (Open-top)
Product Suggestions
Product Suggestions