Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
Supplier: DWK Life Sciences 608122895
Description
- Manufactured from clear 33 expansion borosilicate glass
Specifications
Clear | |
Borosilicate Glass | |
Screw | |
95 mm | |
24-400 |
28 mm | |
40 mL | |
10.82 dr. | |
ASTM Type I, Class A |