Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
Supplier: DWK Life Sciences 608121232
Description
- Manufactured from clear 33 expansion borosilicate glass
Specifications
Clear | |
Borosilicate Glass | |
Screw | |
32 mm | |
8-425 |
12 mm | |
1.8 mL | |
0.5 dr. | |
ASTM Type I, Class A |