Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Packaged in corrugated trays with partitions
Supplier: DWK Life Sciences 608121545
Description
- Manufactured from clear 33 expansion borosilicate glass
Specifications
Clear | |
Borosilicate Glass | |
Screw | |
45 mm | |
13-425 |
15 mm | |
4 mL | |
1 dr. | |
ASTM Type I, Class A |