Learn More
DWK Life Sciences Kimble™ Glass Racks for Westergren Tubes
Reduces the chance of bio hazardous contamination
Manufacturer:  DWK Life Sciences 2215N
 View more versions of this product
Catalog No. 22-295694
Add to cart
Description
Description
  • Racks feature leveling bubbles and adjustment screws
  • Tubes are held firmly in a perpendicular position to ensure accurate readings.
Specifications
Specifications
Not Autoclavable
For westergren and wintrobe tubes
Glass
10
Documents
Accessories