Learn More
DWK Life Sciences Kimble™ Glass Racks for Westergren Tubes
Reduces the chance of bio hazardous contamination
$327.00 - $429.00
Specifications
Autoclavable Not Autoclavable
Material Glass
View More Specs
Products 2
Catalog Number Mfr. No. Positions For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Positions For Use With (Application) Price Quantity & Availability  
22-295693
View Documents
DWK Life Sciences
2212N
6 For westergren tubes
Case for $327.00
Add to cart
22-295694
Accessories View Documents
DWK Life Sciences
2215N
10 For westergren and wintrobe tubes
Case for $429.00
Add to cart
Description
Description
  • Racks feature leveling bubbles and adjustment screws
  • Tubes are held firmly in a perpendicular position to ensure accurate readings.
Specifications
Specifications
Not Autoclavable
Glass