Learn More
Kimble™ KIMAX™ Class B Flasks with standard taper Pennyhead Stopper Available on GSA/VA Contract for Federal Government customers only.
Calibrated “To Contain” within Class B tolerances
Manufacturer:  Kimble™ 28015100
 View more versions of this product
Catalog No. 10-204D
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Calibrated To Contain
13
18cm
100mL
Glass
7.08 in.
±0.16mL
Borosilicate Glass