Learn More
Kimble™ KIMAX™ Colorware™ Flasks
Colorful enamel graduations enhance readability and indicate approximate volumes
Manufacturer:  Kimble™ 26500Y125
 View more versions of this product
Catalog No. 10-200-35
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Rubber
Sunny Yellow Ceramic Enamel scale graduations
66mm
25mL
10.8cm
Matte finish marking
Borosilicate Glass
5
2.59 in.
50 to 125mL
4.25 in.
Narrow
125mL