Kimble™ KIMAX™ Distilling Apparatus Complete Assembly

Complete assembly

Manufacturer: Kimble™ 21500500

 View more versions of this product

Catalog No. 13-631-6

  Check Availability
Add to Cart

    Specifications

    Capacity (English) 16.9 oz.
    Capacity 16.9 oz., 500mL
    Certifications/Compliance USP Type I and ASTM E438, Type I, Class A
    For Use With (Application) Use to distill small amounts of water or in APHA Method 4500-NH3 for determining nitrogen (ammonia) in purified drinking water, natural water and highly purified wastewaters
    Includes 500 or 1000mL flat-bottom flask, distillation head with a 19/38 joint stopper, jacketed graham condenser with 19/38 standard taper top joint
    Standard Taper Joint 19/38
    Standard Taper Stopper No. 22
    Capacity (Metric) 500mL
    Material Borosilicate Glass
    Product Type Distilling Apparatus

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit